C12H316CE7EB Chemcia Scientific, LLC

4-(3-Hydroxy-2-phenyl-propyl)-piperazine-1-carboxylic acid tert-butyl ester

  • MDL number MFCD19441828
  • Cas Number : 189298-13-3
  • Molecular Weight : 320.43546

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=25

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=25
Availability details for
Quantity
  • Availability
  • Add to Cart