C12H316CE7ED Chemcia Scientific, LLC

N-(6-Chloro-pyridazin-3-yl)-acetamide

  • MDL number MFCD09835313
  • Cas Number : 14959-31-0
  • Molecular Weight : 171.59

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name N-(6-chloro-3-pyridazinyl)acetamide
Inchi Code 1S/C6H6ClN3O/c1-4(11)8-6-3-2-5(7)9-10-6/h2-3H,1H3,(H,8,10,11)
Inchi Key UNQTYWWIVXWOJT-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=26

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=26
Availability details for
Quantity
  • Availability
  • Add to Cart