C12H316CE7EF Chemcia Scientific, LLC

2-Cyano-3-methyl-4-(4-nitro-phenyl) -but-2-enoic acid ethyl ester

  • MDL number MFCD19441831
  • Cas Number : 1313714-59-8
  • Molecular Weight : 274.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name ethyl (2E)-2-cyano-3-methyl-4-(4-nitrophenyl)-2-butenoate
Inchi Code 1S/C14H14N2O4/c1-3-20-14(17)13(9-15)10(2)8-11-4-6-12(7-5-11)16(18)19/h4-7H,3,8H2,1-2H3/b13-10+
Inchi Key JCRWSWDLIUAFDG-JLHYYAGUSA-N

Description

Purity 95%
Storage temp. 0~5
Physical Form Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=29

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=29
Availability details for
Quantity
  • Availability
  • Add to Cart