C12H316CE7F4 Chemcia Scientific, LLC

3-(2-Trimethylsilanyl-ethoxymethyl)-6-vinyl-3H-[1,2,3]triazolo[4,5-b]pyridine

  • MDL number MFCD18910942
  • Cas Number : 1313712-65-0
  • Molecular Weight : 276.41655

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=34

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=34
Availability details for
Quantity
  • Availability
  • Add to Cart