C12H316CE7F5 Chemcia Scientific, LLC

4-[2-(2-Trimethylsilanyl-ethoxymethyl)-2H-tetrazol-5-yl]-piperidine-1-carboxylic acid benzyl ester

  • MDL number MFCD19441832
  • Cas Number : 1313712-41-2
  • Molecular Weight : 417.58777

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=35

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=35
Availability details for
Quantity
  • Availability
  • Add to Cart