C12H316CE7F7 Chemcia Scientific, LLC

2-Chloro-3-(4,4,5,5-tetramethyl-[1,3,2]dioxaborolan-2-yl)-phenol

  • MDL number MFCD16994269
  • Cas Number : 1151564-17-8
  • Molecular Weight : 254.52

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-chloro-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenol
Inchi Code 1S/C12H16BClO3/c1-11(2)12(3,4)17-13(16-11)8-6-5-7-9(15)10(8)14/h5-7,15H,1-4H3
Inchi Key DUONOXUXQGHBPZ-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=37

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=37
Availability details for
Quantity
  • Availability
  • Add to Cart