C12H316CE7FB Chemcia Scientific, LLC

N-(4-Bromo-5-methyl-2H-pyrazol-3-yl)-acetamidine

  • MDL number MFCD19441836
  • Cas Number : 1304777-34-1
  • Molecular Weight : 217.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name N-(4-bromo-3-methyl-1H-pyrazol-5-yl)ethanimidamide
Inchi Code 1S/C6H9BrN4/c1-3-5(7)6(11-10-3)9-4(2)8/h2,8H2,1H3,(H2,9,10,11)
Inchi Key WPNBBRTVKICIDB-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. 0~5
Physical Form Off-White Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=53

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=53
Availability details for
Quantity
  • Availability
  • Add to Cart