C12H316CE7FC Chemcia Scientific, LLC

8-Bromo-2,7-dimethyl-3H-pyrazolo[1,5-a][1,3,5]triazin-4-one

  • MDL number MFCD19441837
  • Cas Number : 55457-59-5
  • Molecular Weight : 243.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 8-bromo-2,7-dimethylpyrazolo[1,5-a][1,3,5]triazin-4-ol
Inchi Code 1S/C7H7BrN4O/c1-3-5(8)6-9-4(2)10-7(13)12(6)11-3/h9H,2H2,1H3,(H,10,13)
Inchi Key AKVITCSOLFMCLQ-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=54

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=54
Availability details for
Quantity
  • Availability
  • Add to Cart