C12H316CE804 Chemcia Scientific, LLC

7-Bromo-4-chloro-2,6-dimethyl-thieno[3,2-d]pyrimidine

  • MDL number MFCD19441839
  • Cas Number : 1313712-46-7
  • Molecular Weight : 277.57142

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=60

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=60
Availability details for
Quantity
  • Availability
  • Add to Cart