C12H316CE808 Chemcia Scientific, LLC

4-(3-Chloro-benzyl)-1H-pyrrole-3-carboxylic acid methyl ester

  • MDL number MFCD19441842
  • Cas Number : 1313712-17-2
  • Molecular Weight : 249.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name methyl 4-(3-chlorobenzyl)-1H-pyrrole-3-carboxylate
Inchi Code 1S/C13H12ClNO2/c1-17-13(16)12-8-15-7-10(12)5-9-3-2-4-11(14)6-9/h2-4,6-8,15H,5H2,1H3
Inchi Key CIIWYZBTMMYGHA-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. 0~5
Physical Form Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=63

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=63
Availability details for
Quantity
  • Availability
  • Add to Cart