C12H316CE809 Chemcia Scientific, LLC

4-[2-(4-Fluoro-phenyl)-ethyl]-1H-pyrrole-3-carboxylic acid methyl ester

  • MDL number MFCD19441843
  • Cas Number : 1313712-14-9
  • Molecular Weight : 247.27158

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. 0~5
Physical Form Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=600

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=600
Availability details for
Quantity
  • Availability
  • Add to Cart