C12H316CE80D Chemcia Scientific, LLC

3-Pyridin-3-yl-4H-[1,2,4]oxadiazol-5-one

  • MDL number MFCD12815751
  • Cas Number : 1198-99-8
  • Molecular Weight : 163.1368

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form White Powder
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=68

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=68
Availability details for
Quantity
  • Availability
  • Add to Cart