C12H316CE812 Chemcia Scientific, LLC

5-Bromo-4-chloro-2-fluoro-benzenesulfonyl chloride

  • MDL number MFCD19441846
  • Cas Number : 1070972-67-6
  • Molecular Weight : 307.95404

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=83

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=83
Availability details for
Quantity
  • Availability
  • Add to Cart