C12H316CE814 Chemcia Scientific, LLC

Carbonic acid 4-nitro-phenyl ester 1-chloro-ethyl ester

  • MDL number MFCD19441847
  • Cas Number : 101623-69-2
  • Molecular Weight : 245.62081

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0-5C
Shipping Temperature Ambient
Country of Origin USA
Product Link http://www.chemcia.com/product_fl02.asp?id=89

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=89
Availability details for
Quantity
  • Availability
  • Add to Cart