C12H316CE815 Chemcia Scientific, LLC

(2-Amino-3-methoxy-phenyl)-methanol

  • MDL number MFCD09029933
  • Cas Number : 205877-13-0
  • Molecular Weight : 153.18237

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form Light Yellow Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=95

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=95
Availability details for
Quantity
  • Availability
  • Add to Cart