C12H316CE819 Chemcia Scientific, LLC

N-(5-Fluoro-2-methyl-phenyl)-formamide

  • MDL number MFCD19441849
  • Cas Number : 366-48-3
  • Molecular Weight : 153.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 5-fluoro-2-methylphenylformamide
Inchi Code 1S/C8H8FNO/c1-6-2-3-7(9)4-8(6)10-5-11/h2-5H,1H3,(H,10,11)
Inchi Key PGJLDNYAIHHJJQ-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=103

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=103
Availability details for
Quantity
  • Availability
  • Add to Cart