C12H316CE81A Chemcia Scientific, LLC

N-(3,5-Difluoro-phenyl)-formamide

  • MDL number MFCD00034516
  • Cas Number : 198077-69-9
  • Molecular Weight : 157.1208

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=104

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=104
Availability details for
Quantity
  • Availability
  • Add to Cart