C12H316CE81C Chemcia Scientific, LLC

N-(3-Fluoro-2-methyl-phenyl)-formamide

  • MDL number MFCD19441851
  • Cas Number : 153797-65-0
  • Molecular Weight : 153.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-fluoro-2-methylphenylformamide
Inchi Code 1S/C8H8FNO/c1-6-7(9)3-2-4-8(6)10-5-11/h2-5H,1H3,(H,10,11)
Inchi Key LIAWHEJIUJDVPK-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. 0~5
Physical Form Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=112

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=112
Availability details for
Quantity
  • Availability
  • Add to Cart