C12H316CE81E Chemcia Scientific, LLC

2-Amino-cyclopent-1-enecarboxylic acid ethyl ester

  • MDL number MFCD00216929
  • Cas Number : 7149-18-0
  • Molecular Weight : 155.19831

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form White Needle-Like Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=116

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=116
Availability details for
Quantity
  • Availability
  • Add to Cart