C12H316CE824 Chemcia Scientific, LLC

3-Hydroxy-2-nitro-benzonitrile

  • MDL number MFCD09834347
  • Cas Number : 129298-23-3
  • Molecular Weight : 164.12153

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form Yellow Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=601

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=601
Availability details for
Quantity
  • Availability
  • Add to Cart