C12H316CE829 Chemcia Scientific, LLC

4-Chloro-2,3-dihydro-1H-indole hydrochloride

  • MDL number MFCD09026825
  • Cas Number : 1251023-48-9
  • Molecular Weight : 190.07363

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Salt Data HCl
Storage temp. 0-5*C
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=122

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=122
Availability details for
Quantity
  • Availability
  • Add to Cart