C12H316CE82A Chemcia Scientific, LLC

5-Chloro-2,3-dihydro-1H-indole hydrochloride

  • MDL number MFCD01941330
  • Cas Number : 1013398-58-7
  • Molecular Weight : 190.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 5-chloroindoline hydrochloride
Inchi Code 1S/C8H8ClN.ClH/c9-7-1-2-8-6(5-7)3-4-10-8;/h1-2,5,10H,3-4H2;1H
Inchi Key PUGXFVNHRAMBQK-UHFFFAOYSA-N

Description

Purity 96%
Salt Data HCl
Storage temp. 0~5
Physical Form Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=133

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=133
Availability details for
Quantity
  • Availability
  • Add to Cart