C12H316CE82D Chemcia Scientific, LLC

1-[3-(tert-Butoxycarbonyl-methyl-amino)-propyl]-piperidine-2-carboxylic acid methyl ester

  • MDL number MFCD19441857
  • Cas Number : 1313712-13-8
  • Molecular Weight : 314.43

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name methyl 1-{3-[(tert-butoxycarbonyl)(methyl)amino]propyl}-2-piperidinecarboxylate
Inchi Code 1S/C16H30N2O4/c1-16(2,3)22-15(20)17(4)10-8-12-18-11-7-6-9-13(18)14(19)21-5/h13H,6-12H2,1-5H3
Inchi Key PCKBURMQDNEOFI-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. 0~5
Physical Form Yellow Liquid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=141

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=141
Availability details for
Quantity
  • Availability
  • Add to Cart