C12H316CE82E Chemcia Scientific, LLC

1-[3-(tert-Butoxycarbonyl-methyl-amino)-propyl]-piperidine-4-carboxylic acid ethyl ester

  • MDL number MFCD19441858
  • Cas Number : 1313712-70-7
  • Molecular Weight : 328.45559

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form Red Liquid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=142

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=142
Availability details for
Quantity
  • Availability
  • Add to Cart