C12H316CE83C Chemcia Scientific, LLC

3-(1-Methoxymethyl-butylamino)-propionic acid ethyl ester

  • MDL number MFCD20308330
  • Cas Number : 1343197-89-6
  • Molecular Weight : 217.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name ethyl 3-{[1-(methoxymethyl)butyl]amino}propanoate
Inchi Code 1S/C11H23NO3/c1-4-6-10(9-14-3)12-8-7-11(13)15-5-2/h10,12H,4-9H2,1-3H3
Inchi Key GRQBLEVTPZPHLD-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. 0~5
Physical Form White Crystalline Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=821

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=821
Availability details for
Quantity
  • Availability
  • Add to Cart