C12H316CE856 Chemcia Scientific, LLC

(2,4-Dinitro-phenyl)-acetic acid methyl ester

  • MDL number MFCD00047704
  • Cas Number : 58605-12-2
  • Molecular Weight : 240.17391

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=238

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=238
Availability details for
Quantity
  • Availability
  • Add to Cart