C12H316CE857 Chemcia Scientific, LLC

1-(4-tert-Butyl-phenyl)-piperazine

  • MDL number MFCD01408752
  • Cas Number : 68104-61-0
  • Molecular Weight : 218.34484

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=235

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=235
Availability details for
Quantity
  • Availability
  • Add to Cart