C12H316CE858 Chemcia Scientific, LLC

7H-Pyrrolo[2,3-d]pyrimidin-4-amine sulfuric acid

  • MDL number MFCD09787504
  • Cas Number : 769951-32-8
  • Molecular Weight : 232.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name sulfuric acid compound with 1H-pyrrolo[2,3-d]pyrimidin-4-amine (1:1)
Inchi Code 1S/C6H6N4.H2O4S/c7-5-4-1-2-8-6(4)10-3-9-5;1-5(2,3)4/h1-3H,(H3,7,8,9,10);(H2,1,2,3,4)
Inchi Key ZVFLCRGXDSPVKK-UHFFFAOYSA-N

Description

Purity 97%
Salt Data H2SO4
Storage temp. 0~5
Physical Form Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=212

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=212
Availability details for
Quantity
  • Availability
  • Add to Cart