C12H316CE870 Chemcia Scientific, LLC

5-Methyl-3-(2,2,2-trifluoro-acetylamino)-thiophene-2-carboxylic acid methyl ester

  • MDL number MFCD20921534
  • Cas Number : 1352723-61-5
  • Molecular Weight : 267.22821

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 93
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=811

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=811
Availability details for
Quantity
  • Availability
  • Add to Cart