C12H316CE871 Chemcia Scientific, LLC

3-Nitro-1-(toluene-4-sulfonyl)-1H-pyrrole

  • MDL number MFCD20921535
  • Cas Number : 930111-84-5
  • Molecular Weight : 266.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-[(4-methylphenyl)sulfonyl]-3-nitro-1H-pyrrole
Inchi Code 1S/C11H10N2O4S/c1-9-2-4-11(5-3-9)18(16,17)12-7-6-10(8-12)13(14)15/h2-8H,1H3
Inchi Key PHWKAWLXYVIHMU-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. 0~5
Physical Form Light Brown Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=813

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=813
Availability details for
Quantity
  • Availability
  • Add to Cart