C12H316CE881 Chemcia Scientific, LLC

3-Pyridin-3-yl-morpholine

  • MDL number MFCD04113629
  • Cas Number : 887344-25-4
  • Molecular Weight : 164.20879

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form Yellow Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=861

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=861
Availability details for
Quantity
  • Availability
  • Add to Cart