C12H316CE885 Chemcia Scientific, LLC

6-Bromo-3,4-dihydro-1H-isoquinoline-1,2-dicarboxylic acid 2-tert-butyl ester

  • MDL number MFCD04114851
  • Cas Number : 1260638-11-6
  • Molecular Weight : 356.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 6-bromo-2-(tert-butoxycarbonyl)-1,2,3,4-tetrahydro-1-isoquinolinecarboxylic acid
Inchi Code 1S/C15H18BrNO4/c1-15(2,3)21-14(20)17-7-6-9-8-10(16)4-5-11(9)12(17)13(18)19/h4-5,8,12H,6-7H2,1-3H3,(H,18,19)
Inchi Key QSMPYYITXKRKLS-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=865

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=865
Availability details for
Quantity
  • Availability
  • Add to Cart