C12H316CE88D Chemcia Scientific, LLC

(3,5-Dimethyl-phenyl)-acetonitrile

  • MDL number MFCD00060304
  • Cas Number : 39101-54-7
  • Molecular Weight : 145.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (3,5-dimethylphenyl)acetonitrile
Inchi Code 1S/C10H11N/c1-8-5-9(2)7-10(6-8)3-4-11/h5-7H,3H2,1-2H3
Inchi Key LMUKNQSVBFEUKR-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. 0~5
Physical Form Light Yellow Solid (Low MP)
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=882

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=882
Availability details for
Quantity
  • Availability
  • Add to Cart