C12H316CE891 Chemcia Scientific, LLC

2-Amino-4-(3-bromo-phenyl)-6-(2-methoxy-phenyl)-nicotinonitrile

  • MDL number MFCD04102589
  • Cas Number : 642039-97-2
  • Molecular Weight : 380.24693

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form Off-White to Pale Orange Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=1018

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=1018
Availability details for
Quantity
  • Availability
  • Add to Cart