C12H316CE8A0 Chemcia Scientific, LLC

6-Fluoroimidazo[1,2-a]pyridine-2-carboxylic acid

  • MDL number MFCD07374363
  • Cas Number : 367500-94-5
  • Molecular Weight : 180.13965

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=1061

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=1061
Availability details for
Quantity
  • Availability
  • Add to Cart