C12H316CE8A4 Chemcia Scientific, LLC

5-Bromo-imidazo[1,2-a]pyridine-2-carboxylic acid

  • MDL number MFCD11043994
  • Cas Number : 1026201-52-4
  • Molecular Weight : 241.04525

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=1065

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=1065
Availability details for
Quantity
  • Availability
  • Add to Cart