C12H316CE8A6 Chemcia Scientific, LLC

3-Chloro-5,6,7,8-tetrahydropyrido[4,3-c]pyridazine hydrochloride

  • MDL number MFCD15146561
  • Cas Number : 1314770-27-8
  • Molecular Weight : 206.07588

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. 0~5
Physical Form Brown Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=1174

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=1174
Availability details for
Quantity
  • Availability
  • Add to Cart