C12H316CE8A7 Chemcia Scientific, LLC

{3-[(3-Amino-propyl)-methyl-amino]-propyl}-carbamic acid tert-butyl ester

  • MDL number MFCD22565676
  • Cas Number : 87530-14-1
  • Molecular Weight : 245.36789

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form Light Yellow Liquid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=1217

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=1217
Availability details for
Quantity
  • Availability
  • Add to Cart