C12H316CE8A8 Chemcia Scientific, LLC

6-Benzyl-2,5,6,7-tetrahydro-pyrrolo[3,4-c]pyridazin-3-one

  • MDL number MFCD21086385
  • Cas Number : 1355174-44-5
  • Molecular Weight : 227.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 6-benzyl-2,5,6,7-tetrahydro-3H-pyrrolo[3,4-c]pyridazin-3-one
Inchi Code 1S/C13H13N3O/c17-13-6-11-8-16(9-12(11)14-15-13)7-10-4-2-1-3-5-10/h1-6,9,14H,7-8H2,(H,15,17)
Inchi Key KKDTZYDVAAFRRY-UHFFFAOYSA-N

Description

Purity 92%
Storage temp. 0~5
Physical Form Pink Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=1207

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=1207
Availability details for
Quantity
  • Availability
  • Add to Cart