C12H316CE8A9 Chemcia Scientific, LLC

6-Benzyl-6,7-dihydro-5H-pyrrolo[3,4-c]pyridazin-3-ol

  • MDL number MFCD22565677
  • Cas Number : 1355174-44-5
  • Molecular Weight : 227.26806

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 92
Storage temp. 0~5
Physical Form Pink Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=1208

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=1208
Availability details for
Quantity
  • Availability
  • Add to Cart