C12H316CE8AA Chemcia Scientific, LLC

2-(3-Cydopropoxy-phenyl)-4,4,5,5-tetramethy-[1,3,2]dioxaborolane

  • MDL number MFCD16994464
  • Cas Number : 1035690-24-4
  • Molecular Weight : 260.14382

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. 0~5
Physical Form Pale Yellow Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=1182

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=1182
Availability details for
Quantity
  • Availability
  • Add to Cart