C12H316CE8AE Chemcia Scientific, LLC

3-Amino-5-bromo-isonicotinic acid trifluoroacetate

  • MDL number MFCD26792513
  • Cas Number : 1951441-00-1
  • Molecular Weight : 331.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2,2,2-trifluoroacetic acid compound with 3-amino-5-bromoisonicotinic acid (1:1)
Inchi Code 1S/C6H5BrN2O2.C2HF3O2/c7-3-1-9-2-4(8)5(3)6(10)11;3-2(4,5)1(6)7/h1-2H,8H2,(H,10,11);(H,6,7)
Inchi Key CJFHVXDKZSAIPT-UHFFFAOYSA-N

Description

Purity 95%
Salt Data TFA
Storage temp. 0~5
Physical Form Green Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=2025

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=2025
Availability details for
Quantity
  • Availability
  • Add to Cart