C12H316CE8B2 Chemcia Scientific, LLC

(R)-4-(8-Cyclopentyl-7-ethyl-5-methyl-6-oxo-5,6,7,8-tetrahydro-pteridin-2-ylamino) -3-methoxy-benzoic acid

  • MDL number MFCD18207100
  • Cas Number : 755039-56-6
  • Molecular Weight : 425.49159

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. 0~5
Physical Form Off-White Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=1475

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=1475
Availability details for
Quantity
  • Availability
  • Add to Cart