C12H316CE8B3 Chemcia Scientific, LLC

(R)-2-Chloro-8-cyclopentyl-7-ethyl-7,8-dihydro-5H-pteridin-6-one

  • MDL number MFCD11977211
  • Cas Number : 755039-54-4
  • Molecular Weight : 280.75964

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form Off-White Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=1477

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=1477
Availability details for
Quantity
  • Availability
  • Add to Cart