C12H316CE8B6 Chemcia Scientific, LLC

4-Chloro-5,6,7,8-tetrahydro-quinazolin-2-ylamine

  • MDL number MFCD13689131
  • Cas Number : 111896-77-6
  • Molecular Weight : 183.64

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-chloro-5,6,7,8-tetrahydro-2-quinazolinamine
Inchi Code 1S/C8H10ClN3/c9-7-5-3-1-2-4-6(5)11-8(10)12-7/h1-4H2,(H2,10,11,12)
Inchi Key BTLNLOHFQGIGLV-UHFFFAOYSA-N

Description

Purity 98%
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=1481

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=1481
Availability details for
Quantity
  • Availability
  • Add to Cart