C12H316CE8B9 Chemcia Scientific, LLC

2-Amino-5,8-dihydro-6H-pyrido[3,4-d]pyrimidine-7-carboxylic acid tert-butyl ester

  • MDL number MFCD22565681
  • Cas Number : 1395079-01-2
  • Molecular Weight : 250.30286

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=1485

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=1485
Availability details for
Quantity
  • Availability
  • Add to Cart