C12H316CE8BC Chemcia Scientific, LLC

6,7-Dihydro-5H-pyrrolo[3,4-d]pyrimidin-2-ylamine dihydrochloride

  • MDL number MFCD08457553
  • Cas Number : 157327-52-1
  • Molecular Weight : 209.0794

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. 0~5
Physical Form Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=1488

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=1488
Availability details for
Quantity
  • Availability
  • Add to Cart