C12H316CE8C0 Chemcia Scientific, LLC

4,5,6,7-Tetrahydro-2H-pyrazolo[4,3-c]pyridine dihydrochloride

  • MDL number MFCD13368261
  • Cas Number : 410544-19-3
  • Molecular Weight : 196.08067

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 92
Salt Data 2HCl
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=1494

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=1494
Availability details for
Quantity
  • Availability
  • Add to Cart