C12H316CE8C2 Chemcia Scientific, LLC

4,5,6,7-Tetrahydro-2H-pyrazolo[3,4-c]pyridine dihydrochloride

  • MDL number MFCD22570200
  • Cas Number : 871726-74-8
  • Molecular Weight : 196.08067

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. 0~5
Physical Form Off-White Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=1496

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=1496
Availability details for
Quantity
  • Availability
  • Add to Cart