C12H316CE8C5 Chemcia Scientific, LLC

2-(4-Fluoro-phenyl)-6,7-dihydro-5H-pyrrolo[3,4-d]pyrimidine hydrochloride

  • MDL number MFCD22570195
  • Cas Number : 1951441-05-6
  • Molecular Weight : 251.69297

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. 0~5
Physical Form Off-White Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=1509

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=1509
Availability details for
Quantity
  • Availability
  • Add to Cart